Order Entry
Canada
ContactUsLinkComponent
Scandium ICP Standard, 1 ml = 1 mg Sc (1,000 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Scandium ICP Standard, 1 ml = 1 mg Sc (1,000 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCPSC1KN-500
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Scandium ICP Standard, 1 ml = 1 mg Sc (1,000 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCPSC1KN-500
Supplier:  Ricca Chemical
Supplier Number:  RPSC1KN0-500N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    3% HNO₃
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. no.:
    RCPSC1KN-500
  • Element:
    Scandium
  • Supplier No.:
    RPSC1KN0-500N

Specifications

About this item