Order Entry
Canada
ContactUsLinkComponent
 
Lanthanum ICP Standard, 1 ml = 10 mg La (10000 ppm La)La₂O₃ in 5% HNO₃, Ricca Chemical Company
  RCPLA10KN-100
Small Business Enterprise
 :  
undefined
Lanthanum ICP Standard, 1 ml = 10 mg La (10000 ppm La)La₂O₃ in 5% HNO₃, Ricca Chemical Company
  RCPLA10KN-100
 :  RPLA10KN-100N
Small Business Enterprise
 :  

 

  • Concentration:
    10000 µg/ml
  • Matrix:
    5% HNO₃
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. no.:
    RCPLA10KN-100
  • Element:
    Lanthanum
  • Supplier No.:
    RPLA10KN-100N