Lanthanum ICP Standard, 1 ml = 1 mg La (1000 ppm La)La₂O₃ in 3% HNO₃, Ricca Chemical Company
RCPLA1KN-100
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. no.:RCPLA1KN-100
- Element:Lanthanum
- Supplier No.:RPLA1KN0-100N