Iridium ICP Standard, 1 ml = 1 mg Ir (1000 ppm Ir)IrCl₃ in 10% HCl, Ricca Chemical Company
RCPIR1KH-500
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:10% HCl
- Pack type:Poly
- Size:500 ml
- Cat. no.:RCPIR1KH-500
- Element:Iridium
- Supplier No.:RPIR1KH0-500N