Calcium Standard Stock Solution (1000 ppm Ca), Ricca Chemical Company
RC1821-16
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:H₂O/tr. AcOH
- Pack type:Poly Bottle
- Size:500 ml
- Cat. no.:RC1821-16
- Element:Calcium
- Supplier No.:R1821000-500A