Zirconium ICP Standard, 1 ml = 10 mg Zr (10000 ppm Zr)ZrO(NO₃)₂ in 5% HNO₃, RICCA Chemical Company
RCPZR10KN-100
Small Business Enterprise
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Concentration:10000 µg/ml
- Matrix:5% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. no.:RCPZR10KN-100
- Element:Zirconium
- Supplier No.:RPZR10KN-100N