Mercury GFAA Cal / Spike Standard - 100 ppm Hg in 5% HNO₃, RICCA Chemical Company
Catalog # RCRGFAAHG-50
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:100 µg/ml
- Matrix:5% HNO₃
- Pack type:Poly
- Size:50 ml
- Cat. no.:RCRGFAAHG-50
- Element:Mercury
- Supplier No.:RGFAAHG0-50N