Nickel AA Standard, 1 ml = 1 mg Ni (1000 ppm Ni)Ni in 3% HNO₃, RICCA Chemical Company
Catalog # RCANI1KN-500
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:500 ml
- Cat. no.:RCANI1KN-500
- Element:Nickel
- Supplier No.:RANI1KN0-500N