Order Entry
Canada
ContactUsLinkComponent
 
Indium AA Standard, 1,000 ppm in 3% HNO₃, Ricca Chemical Company
  RCAIN1KN-500
Small Business Enterprise
 :  
undefined
Indium AA Standard, 1,000 ppm in 3% HNO₃, Ricca Chemical Company
  RCAIN1KN-500
 :  RAIN1KN0-500N
Small Business Enterprise
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    3% HNO₃
  • Pack type:
    Poly
  • Size:
    500 ml
  • Cat. no.:
    RCAIN1KN-500
  • Element:
    Indium
  • Supplier No.:
    RAIN1KN0-500N