Order Entry
Canada
ContactUsLinkComponent
 
Gallium ICP Standard, 1 ml = 1 mg Ga (1000 ppm Ga)Ga in 3% HNO₃, Ricca Chemical Company
  RCPGA1KN-100
Small Business Enterprise
 :  
undefined
Gallium ICP Standard, 1 ml = 1 mg Ga (1000 ppm Ga)Ga in 3% HNO₃, Ricca Chemical Company
  RCPGA1KN-100
 :  RPGA1KN0-100N
Small Business Enterprise
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    3% HNO₃
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. no.:
    RCPGA1KN-100
  • Element:
    Gallium
  • Supplier No.:
    RPGA1KN0-100N