Manganese ICP Standard, 1 ml = 1 mg Mn (1000 ppm Mn)Mn(C₂H₃O₂)₂ in 3% HNO₃, RICCA Chemical Company
Catalog # RCPMN1KN-500
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:500 ml
- Cat. no.:RCPMN1KN-500
- Element:Manganese
- Supplier No.:RPMN1KN0-500N