Order Entry
Canada
ContactUsLinkComponent
 
Cesium ICP Standard, 1 ml = 10 mg Cs (10,000 ppm Cs)CsNO₃ in 5% HNO₃, Ricca Chemical Company
  RCPCS10KN-500
Small Business Enterprise
 :  
undefined
Cesium ICP Standard, 1 ml = 10 mg Cs (10,000 ppm Cs)CsNO₃ in 5% HNO₃, Ricca Chemical Company
  RCPCS10KN-500
 :  RPCS10KN-500N
Small Business Enterprise
 :  

 

  • Concentration:
    10000 µg/ml
  • Matrix:
    5% HNO₃
  • Pack type:
    Poly
  • Size:
    500 ml
  • Cat. no.:
    RCPCS10KN-500
  • Element:
    Cesium
  • Supplier No.:
    RPCS10KN-500N