Order Entry
Canada
ContactUsLinkComponent
Cesium AA Standard, 1 ml = 1 mg Cs (1,000 ppm Cs)CsNO₃ in 3% HNO₃, Ricca Chemical Company
Cesium AA Standard, 1 ml = 1 mg Cs (1,000 ppm Cs)CsNO₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCACS1KN-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Cesium AA Standard, 1 ml = 1 mg Cs (1,000 ppm Cs)CsNO₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCACS1KN-100
Supplier:  Ricca Chemical
Supplier Number:  RACS1KN0-100N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    3% HNO₃
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. no.:
    RCACS1KN-100
  • Element:
    Cesium
  • Supplier No.:
    RACS1KN0-100N

Specifications

About this item