You searched for: Centrifuge Tubes
These disposable conical-bottom and freestanding tubes feature easy-to-read graduations and smooth inner walls for easy filling and sample preparation. Rack-packed tubes are packaged in freezable, recyclable, autoclavable racks, and stackable rack designs make them ideal for benchtop organization. Caps are available in either flat or plug styles. Certain models are designed to provide protection for light-sensitive samples, manufactured from a dark polypropylene. These high-quality, high-performing centrifuge tubes are ideal for maintaining a secure seal when using shakers and horizontal incubators.
Nalgene® Conical Oak Ridge Centrifuge Tube, Graduated, PPCO, Thermo Scientific
Supplier: Thermo Fisher Scientific
These conical centrifuge tubes combine the better separation/pelleting capabilities of a conical tube with the superior sealing capability of Oak Ridge tubes.
Expand 1 Items
Flip-Top Centrifuge Tubes, Argos Technologies
Supplier: Argos Technologies
The Argos flip-top centrifuge tube offers a hinged flip-top cap that allows for single-handed opening and closing.
Expand 1 Items
KIMAX® Oil and Weathering/End Point Index Centrifuge Tube, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Designed for use in the determination of residues in liquified petroleum (LP) gases.
Expand 1 Items
Nalgene® Oak Ridge Centrifuge Tubes, Polypropylene Copolymer, Thermo Scientific
Supplier: Thermo Fisher Scientific
Contact-clear, leakproof polypropylene copolymer tubes allow easy examination of contents.
Expand 8 Items
Nunc® Conical Centrifuge Tube, 250 ml, Thermo Scientific
Supplier: Thermo Fisher Scientific
The Nunc® 250 ml conical centrifuge Tube offers added capacity for larger samples with a one-of-a-kind wide mouth design.
Expand 1 Items
KIMBLE® CaliFornia Centrifuge Tube, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Manufactured from borosilicate glass 3.3. Tube has a permanent red stripe under the white enamel graduations for easy reading of results.
Expand 1 Items
V-bottom Clear Tubes with Caps, 2.0 ml, Zymo Research
Supplier: Zymo Research
Clear 2.0 ml skirted tubes made of durable polypropylene construction.
Expand 2 Items
SP Wilmad-LabGlass Capillary Tip Centrifuge Tube, SP Industries
Supplier: SP Industries
Designed for use in ASTM D-2273 and ASTM D-2709, this borosilicate glass capillary tipped long form centrifuge tube is 100 ml in length.
Expand 1 Items
KIMAX® Oil Centrifuge Tube, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Designed to accommodate a wide variety of applications.
Expand 1 Items
KIMAX® Centrifuge Tubes with [ST] Glass Stopper, Conical-Bottom, Graduated, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Tubes feature solid, pennyhead [ST] stoppers.
Expand 2 Items
Nalgene® Round-Bottom Centrifuge Tubes, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
Strong, crystal clear polycarbonate tubes for use with temperatures from –135 to 135 °C (–211 to 275 °F).
Expand 7 Items
Kimble® Disposable Plain Conical Centrifuge Tubes with Snap Cap Rim, Glass, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Borosilicate glass (N-51A) conical tubes are constructed with a finished top for polyethylene snap-top closures.
Expand 4 Items
Plain Reusable Centrifuge Tubes with Screw Caps, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Threaded ungraduated conical centrifuge tube with a PTFE/rubber-lined black phenolic screw cap.
Expand 1 Items
Nunc® Centrifuge Tubes, Polypropylene, Conical, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
The Nunc® conical sterile polypropylene centrifuge tubes are premium, high quality, and environmentally friendly conical tubes.
Expand 4 Items
KIMAX® Centrifuge Tubes, Graduated, Oil, with White Scales, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements and it is used in the determination of bottom sediment and water in petroleum products.
Expand 1 Items
U-bottom Clear Tubes with Caps, 2.0 ml, Zymo Research
Supplier: Zymo Research
Clear 2.0 ml skirted tubes made of durable polypropylene construction.
Expand 2 Items
SP Wilmad-LabGlass Bulb Oil Centrifuge Tubes, SP Industries
Supplier: SP Industries
Calibrated 'to contain' (TC), these Goetz-type borosilicate glass bulb oil centrifuge tubes are pear shaped, similar to LG-4420, but they have a lower stem calibrated to hold 1.5 ml.
Expand 1 Items
Nalgene® Oak Ridge Centrifuge Tubes, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
Transparent autoclavable polycarbonate tubes.
Expand 8 Items
Nalgene® Oak Ridge Centrifuge Tubes, Teflon® FEP, Thermo Scientific
Supplier: Thermo Fisher Scientific
For high-speed centrifugation of aggressive chemicals.
Expand 3 Items
KIMAX® Graduated Hopkins Vaccine Centrifuge Tube, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
KIMAX® tube is designed to standardize vaccines.
Expand 1 Items
KIMAX® Centrifuge Tube, Conical-Bottom, Graduated, with Pourout, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Tube has a short tapered bottom and a pour spout.
Expand 1 Items
Nalgene® Centrifuge Tubes, Polypropylene Copolymer (PPCO), Conical-Bottom, Graduated, Thermo Scientific
Supplier: Thermo Fisher Scientific
These tubes offer excellent chemical resistance.
Expand 2 Items
Nalgene® Oak Ridge Conical Centrifuge Tube Adapter; white PC, Thermo Scientific
Supplier: Thermo Fisher Scientific
Nalgene oak ridge conical tube adapter must be used with Nalgene 35 L conical oak ridge tubes.
Expand 1 Items
U-bottom Amber Tubes with Caps, 2.0 ml, Zymo Research
Supplier: Zymo Research
Amber 2.0 ml skirted tubes made of durable polypropylene construction.
Expand 2 Items
V-bottom Amber Tubes with Caps, 2.0 ml, Zymo Research
Supplier: Zymo Research
Amber 2.0 ml skirted tubes made of durable polypropylene construction.
Expand 2 Items
Color Stability Tube, Ace Glass
Supplier: Ace Glass
Used in tentative AOCS Method L-15A-58 for measuring the processing color stability of fatty acids.
Expand 3 Items
SP Bel-Art CUBE 2UBE™ Rack and Tubes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
CUBE 2UBE™ Tubes offer the advantages and convenience of pipetting 2.2 ml per tube in an 8×12 grid, 96-well format.
Expand 3 Items
Sample Preparation, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Agilent offers a range of sample preparation products to support your workflows.
Expand 1 Items
Nunc® Disposable Tubes, Round-Bottom, Polystyrene, Thermo Scientific
Supplier: Thermo Fisher Scientific
These transparent, polystyrene tubes offer a high degree of thermal and chemical stability.
Expand 3 Items
Quick-Seal Ultracentrifugation Tubes, Beckman Coulter
Supplier: Beckman Coulter
Quick-seal tubes eliminate most of the steps involved in capping tubes and cut handling time in half.