HyClone AdvanceSTEM™ Stem Cell Media, Mesenchymal Stem Cell Expansion Kit, HyClone products (Cytiva)
Supplier: Cytiva
Certificates
About this item
AdvanceSTEM™ Mesenchymal Stem Cell Basal Medium and Mesenchymal Stem Cell Growth Supplement have been formulated to support expansion and maintenance of undifferentiated human mesenchymal stem cells (hMSCs).
- Mycoplasma free and sterility tested
- Stored at –10 °C or lower away from light
Supported cells include Human Wharton's Jelly Stem Cells (HWJSCs), Human Adipose-derived Mesenchymal Stem Cells (HAMSCs), Human Placental Mesenchymal Stem Cells (HPMSCs), Multipotent Cord Blood Unrestricted Somatic Stem Cells (MUCBUSSCs) and bone marrow-derived hMSCs.
It provides optimized conditions for embryonic stem cell expansion and differentiation.
Specifications
- Storage temperature:Less than –10 °C
Specifications
Related Information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together