Orders LinkContactUsLinkComponent
Hydrion® Sanitizer Test Kits, Micro Essential
Supplier:   Micro Essential
Hydrion® Sanitizer Test Kits, Micro Essential
Supplier:  Micro Essential
Certificates
Hydrion® Sanitizer Test Kits, Micro Essential
Hydrion® Sanitizer Test Kits, Micro Essential
4 Options Available Below
Product Details & Documents
Use Hydrion Sanitizer test papers wherever food is commercially produced or served to test the concentration of sanitizers, as required by health code
Ammonium test kit and kit refill are for testing first and second generation Quats. Match points at 0, 100, 200, 300, and 400ppm.
Chlorine test kit and kit refill are for testing free chlorine in sanitizing rinse. Match points at 0, 50 100, and 200ppm.
These test papers provide a simple, reliable way to meet Federal, State, and local food safety guidelines
Product Family Options
Product Information
  • Range
    Pack type
    Availability
    Price
  • 60720-300
    QT-10 Quaternary Ammonium Test Kit, 0-400 ppm
    QT-10 Quaternary Ammonium Test Kit, 0-400 ppm
    0-400 ppm
    Contains dispenser, color chart and 1 roll of test paper, enough for 100 tests or more.
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $23.69
    $23.69
    Log in for availability
    Case of 10
    Case of 10 - $236.90
    $236.90
    Each
       $23.69
    Case of 10
       $236.90
    Supplier Number: QT-10
  • 60803-580
    CM-240 Chlorine Test Kit, 0-200 ppm
    CM-240 Chlorine Test Kit, 0-200 ppm
    0-200 ppm
    Contains dispenser, color chart and 2 rolls of test paper, enough for 200 tests or more.
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $14.44
    $14.44
    Log in for availability
    Case of 10
    Case of 10 - $144.40
    $144.40
    Each
       $14.44
    Case of 10
       $144.40
    Supplier Number: CM-240
  • 60803-604
    Chlorine Test Kit Refill
    Chlorine Test Kit Refill
    Contains 10 packs of 5 rolls each
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $28.23
    $28.23
    Log in for availability
    Case of 50
    Case of 50 - $282.30
    $282.30
    Pack of 5
       $28.23
    Case of 50
       $282.30
    Supplier Number: CMR-245
  • 34175-496
    Quaternary Ammonium Kit Refill
    Quaternary Ammonium Kit Refill
    Contains 10 packs of 5 rolls each.
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 5 (1/Itm/Bx)
    Pack of 5 (1/Itm/Bx) - $84.82
    $84.82
    Log in for availability
    Case of 50 (1/ Itm/Bx)
    Case of 50 (1/ Itm/Bx) - $848.20
    $848.20
    Pack of 5 (1/Itm/Bx)
       $84.82
    Case of 50 (1/ Itm/Bx)
       $848.20
    Supplier Number: QTR-10