Specifications
- Description:LabTower TII UV 40 LPH System, 40 L/hr output
- Environmentally Preferable:
- Production flow rate:40 L/h
- W×D×H:450×580×1500 mm (17.7×22.8×59.1")
- Cat. no.:89230-412
- Supplier No.:50132141
Specifications
About this item
The Barnstead LabTower TII water purification systems provides up to 200L/day of Type 2 (10-15 megohm) water from tap water
The LabTower TII systems are completely free-standing units which require no bench or wall space. The unit comes standard with casters for easy moving.
The LabTower TII systems are suited to meet a wide range of applications needs. Type 2 water can be used for rinsing glassware, supplying water to autoclaves, waterbaths, or Type 1 systems that require pretreated feed water and preparing or diluting buffers, reagents or media for example.
All LabTower TII systems include: quick discconnects to make changing consumables easy, built in 100L reservoir and all core consumables and tubing. Standard with a 100L integrated reservoir.