You searched for: Heating Mantles
When handling organic liquid samples, the heating mantles are alternatives to Bunsen burners and hotplates. In contrast to other heating devices, glassware containers may be placed in direct contact with the heating mantle without the risk of glassware shattering. While fabric constructed styles can be molded to accommodate a wide variety of flask shapes and sizes, the rigid heating mantles are less susceptible to spill or tearing damage.
Glas-Col® Aluminum Housing Heating Mantle, Ace Glass
Supplier: Ace Glass
This mantle comes with a hole in bottom to accommodate flasks with bottom outlets
Expand 8 Items
SP Wilmad-LabGlass Hemispherical Heating Mantles, SP Industries
Supplier: SP Industries
Made of soft, all glass fabric that supports the bottom half of a flask, these hemispherical heating mantles allow for full view of the contents of single or multi-neck flasks
Expand 10 Items
SP Wilmad-LabGlass Aluminum Bench Top Hemi Heating Mantles, SP Industries
Supplier: SP Industries
These benchtop heating mantles are used for round bottom glass flasks.
Expand 6 Items
Mantle, Aluminum Housing, Ace Glass Incorporated
Supplier: Ace Glass
Resin and reaction flask mantle
Expand 3 Items
Glas-Col® Heating Mantle, Ace Glass
Supplier: Ace Glass
Fabric heating mantle fits reaction flasks ranging from 100mL to 6000mL, including Ace Scale-Up Series flasks.
Expand 8 Items
SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Supplier: SP Industries
Designed for use with cylindrical reaction kettles LG-8071 through LG-8082, these heating mantles have an aluminum housing exterior.
Expand 2 Items
Glas-Col® Fabric Insulating Tops, Ace Glass
Supplier: Ace Glass
Fabric insulating tops for 1, 2, and 3 neck flasks. Lacing closure allows installation without disturbing attached glassware or equipment. Insulating tops have no electrical connections.
Expand 6 Items
Heating Mantles, for Hydrogenation/Gas Bottles, Ace Glass Incorporated
Supplier: Ace Glass
These heating mantles are designed specifically for use with Hydrogenation/Gas bottle sizes 250mL (89058-840), 500mL (89058-842), 1000mL (89058-844), and 2500mL (89058-846).
Expand 1 Items
Mantle, Aluminum Housing, Bottom Opening, Ace Glass Incorporated
Supplier: Ace Glass
Aluminum housed heating mantles feature a bottom opening to accommodate flasks with bottom valves
Expand 8 Items
SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Supplier: SP Industries
These all-glass constructed heating mantles are for use with LG-8071 through LG-8089 reaction kettles. They provide even heat distribution.
Expand 1 Items
Mantle, Aluminum Housing, Solid Bottom, Ace Glass Incorporated
Supplier: Ace Glass
Solid, aluminum-housed heating mantles are versatile enough to be used on lab benches, stands, or floors
Expand 9 Items
Mantle for Griffin-Type Beakers, Ace Glass Incorporated
Supplier: Ace Glass
Heating mantles designed for use with Griffin-type beakers
Expand 11 Items
Flask Heater, Ace Glass Incorporated
Supplier: Ace Glass
Specially designed glass/fiber cloth mantle with bottom opening and zipper for use with flasks to allow valve to protrude below heater
Expand 4 Items
Glas-Col® Flexible Fabric Heating Mantle, Antylia Scientific
Supplier: Argos Technologies
Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes.
Expand 13 Items
Heating Mantle, Pear Shaped, Ace Glass Incorporated
Supplier: Ace Glass
Heating Mantle for use with pear-shaped flasks
Expand 5 Items
Heating Mantle Micro, Glas-Col®
Supplier: Glas-Col
Precise heat control for small flasks.
Expand 3 Items
Glas-Col® STM Series Mantles, High Temperature, Chemglass
Supplier: Chemglass
The Glas-Col® Series STM mantle performs the same function as a Series TM heating mantle, plus operations requiring more heat.
Expand 6 Items
Uncontrolled Electromantles, with Metal Case, Electrothermal
Supplier: Argos Technologies
CMU heating mantles are used in wet chemistry to heat liquids in round bottom flasks
Expand 1 Items
Mantles, Fabric/Cloth, Ace Glass Incorporated
Supplier: Ace Glass
Heating mantles are designed for use with reaction flasks
Expand 5 Items
Glas-Col® Series M Spherical Heating Mantles, Chemglass
Supplier: Chemglass
The Glas-Col® Series M mantle is the most compact mantle, with an aluminum shell that can heat flasks up to 72 liters.
Expand 7 Items
Sand Bath Heating Mantles, DWK Life Sciences
Supplier: Kimble Chase
Sand bath heating mantle is electrically heated and utilizes commercially available sand (16 to 30 mesh).
Expand 1 Items
Heating Mantles, Upper Portion for Unjacketed Cylindrical Vessels, Chemglass
Supplier: Chemglass
Glass fabric heating mantle with a lengthwise slit, held together with a hook and lace closure.
Expand 6 Items
Electric Heating Mantles
Supplier: Glas-Col
Designed to Operate Below Red Heat at Maximum Input Voltage.
Expand 3 Items
Heating Mantles, Griffin Beakers, Glas-Col®
Supplier: Glas-Col
Excellent fit and heating for griffin beakers.
Expand 7 Items
Heating Mantles, Spherical Flask, 230 V CE, Glas-Col®
Supplier: Glas-Col
Excellent fit and heating for spherical flasks.
Expand 1 Items
Heating Mantles, Lower Portions, Unjacketed Cylindrical Vessels, Chemglass
Supplier: Chemglass
For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels.
Expand 4 Items
Heating Mantle, Buchner Funnel, Glas-Col®
Supplier: Glas-Col
Heating for buchner funnel mantles.
Expand 4 Items
Heating Mantle, Gallon Can, Glas-Col®
Supplier: Glas-Col
Mantle for gallon can.
Expand 1 Items
Resin and Reaction Flask Mantle, Aluminum Housing, Bench-Top, Ace Glass Incorporated
Supplier: Ace Glass
Resin and reaction flask mantle is bench-top size with a small footprint
Expand 5 Items
Glas-Col® Heating Mantle Tops, Chemglass
Supplier: Chemglass
These Glas-Col® mantles help prevent vapor condensation in the upper half of a flask during distillation of high-boiling materials.