ContactUsLinkComponent
39 results for Microplates
You searched for: Solid Phase Extraction Plates (SPE)
Solid Phase Extraction Plates (SPE)
Solid phase extraction plates are ideal for SPE sample preparing where dissolved or suspended compounds are separated from others in a liquid mixture. Designed specifically for bioanalytical and clinical elution methods, the one-piece, frit-less plates have the highest throughput workflow for the most complete phase partition. The solid phase extraction plates have optimum testing reproducibility and cleanliness when removing semivolatile or nonvolatile analytes and solid samples.
Sort by
J.T.Baker® BAKERBOND® SPE 96-Well Plates
J.T.Baker® BAKERBOND® SPE 96-Well Plates
Supplier: HICHROM LIMITED
Solid Phase Extraction (SPE) is commonly used as a highly selective sample clean-up or sample enrichment method prior to subsequent analyses. J.T.Baker® BAKERBOND® SPE 96 well plates are well suited for high throughput and automated SPE methods. With complementary high capacity sorbents to existing spe extraction columns in the J.T.Baker® BAKERBOND® range, higher throughput methodologies can now be simply developed and automated.
Expand 24 Items
Loading...
HyperSep™ C18 SPE Plates
HyperSep™ C18 SPE Plates
Supplier: Thermo Scientific
Feature a highly retentive alkyl-bonded phase for non-polar to moderately polar compounds. Silica material used for packaging.
Expand 4 Items
Loading...
HyperSep™ Hypercarb SPE Plates
HyperSep™ Hypercarb SPE Plates
Supplier: Thermo Scientific
Unique material for retention of highly polar compounds.
Expand 3 Items
Loading...
HyperSep™ Retain AX SPE Plates
HyperSep™ Retain AX SPE Plates
Supplier: Thermo Scientific
Versatile polymeric material for retention of acidic compounds.
Expand 1 Items
Loading...
HyperSep™ SAX Strong Anion Exchanger SPE Plates
HyperSep™ SAX Strong Anion Exchanger SPE Plates
Supplier: Thermo Scientific
Strong anion exchange sorbent for extraction of weak acids.
Expand 1 Items
Loading...
HyperSep™ SCX Strong Cation Exchanger SPE Plates
HyperSep™ SCX Strong Cation Exchanger SPE Plates
Supplier: Thermo Scientific
A strong cation sorbent for extraction of charged basic compounds.
Expand 2 Items
Loading...
HyperSep™ Silica SPE Plates
HyperSep™ Silica SPE Plates
Supplier: Thermo Scientific
A polar sorbent primarily used to retain analytes in non-polar matrices.
Expand 4 Items
Loading...
HyperSep™ Diol SPE Plates
HyperSep™ Diol SPE Plates
Supplier: Thermo Scientific
A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.
Expand 4 Items
Loading...
HyperSep™ Phenyl SPE Plates
HyperSep™ Phenyl SPE Plates
Supplier: Thermo Scientific
HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation.
Expand 1 Items
Loading...
HyperSep™ Protein Precipitation Plate, Thermo Scientific
Supplier: Thermo Scientific
HyperSep™ Protein Precipitation Plates provide a quick, effective approach for removal of proteins from biological compounds using the protein crash technique.
Expand 1 Items
Loading...
HyperSep™ Cyano SPE Plates
HyperSep™ Cyano SPE Plates
Supplier: Thermo Scientific
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation.
Expand 4 Items
Loading...
HyperSep™ Retain PEP SPE Plates
HyperSep™ Retain PEP SPE Plates
Supplier: Thermo Scientific
Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation. Versatile polymeric material for retention of polar and non-polar analytes.
Expand 2 Items
Loading...
SOLA Solid Phase Extraction Plates and Cartridges, Thermo Scientific
SOLA Solid Phase Extraction Plates and Cartridges, Thermo Scientific
Supplier: Thermo Scientific
SOLA is a state of the art fritless polymeric SPE that reduces, channeling, voiding, packing inconsistencies and reduces the need for expense retesting of critical samples.
Expand 15 Items
Loading...
HyperSep™ Aminopropyl SPE Plates
HyperSep™ Aminopropyl SPE Plates
Supplier: Thermo Scientific
A polar sorbent for both polar and anion exchange interactions.
Expand 3 Items
Loading...
HyperSep™ C8 SPE Plates
HyperSep™ C8 SPE Plates
Supplier: Thermo Scientific
HyperSep C8 SPE well plates and individual wells provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.
Expand 4 Items
Loading...
SPEC Disk SPE Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Plate, 96-well, Bond Elut SPEC DAS
Expand 8 Items
Loading...
Bond Elut™ 96 Square-Well Plates, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Bond Elut 96 square-well plates support well-plate SPE applications with larger sample volumes.
Expand 3 Items
Loading...
Cleanert® CN SPE Plates, 96-Well, Bonna Agela
Cleanert® CN SPE Plates, 96-Well, Bonna Agela
Supplier: Agela Technologies
These 96-well plates have a particle size of 40 to 60 µm and a pore size of 60 Å.
Expand 4 Items
Loading...
Empore™ 96-Well Solid Phase Extraction Plates, CDS Analytical
Empore™ 96-Well Solid Phase Extraction Plates, CDS Analytical
Supplier: CDS ANALYTICAL
Empore SPE 96-well plates improve data and throughput at a low cost.
Expand 5 Items
Loading...
Kinesis® TELOS® neo™ MicroPlate™ SPE Populated 96-Well Microplates, Cole-Parmer®
Kinesis® TELOS® neo™ MicroPlate™ SPE Populated 96-Well Microplates, Cole-Parmer®
Supplier: Cole-Parmer Instrument Co.
Elution volume, 50 µl for minimal sample dilution and reduced evaporation time.
Expand 10 Items
Loading...
Kinesis® TELOS® neo™ SPE Fixed 96-Well Microplates, Cole-Parmer®
Kinesis® TELOS® neo™ SPE Fixed 96-Well Microplates, Cole-Parmer®
Supplier: Cole-Parmer Instrument Co.
Modular design lets you simply remove any unwanted wells and add sealing strips.
Expand 9 Items
Loading...
Kinesis® TELOS® neo™ MicroPlate™ SPE Loose 96-Well Microplates, Cole-Parmer®
Kinesis® TELOS® neo™ MicroPlate™ SPE Loose 96-Well Microplates, Cole-Parmer®
Supplier: Cole-Parmer Instrument Co.
Populate base plate with only the wells needed.
Expand 9 Items
Loading...
Bond Elut SI Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut SI is particularly effective at separating compounds with a very similar structure.
Expand 1 Items
Loading...
Bond Elut Lipid Extraction Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Bond Elut Lipid Extraction SPE 1 mL cartridges and 2 mL 96-well plates provide highly selective and efficient extraction of lipids for lipid analysis, lipid profiling, and lipidomics workflows.
Expand 1 Items
Loading...
VersaPlate Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Versa plate allows you to customize with different numbers of samples or sorbent phase types.
Expand 1 Items
Loading...
Bond Elut C18 EWP Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Bond Elut C18 EWP features wide pores, making it ideal for efficient extraction of macromolecules.
Expand 2 Items
Loading...
Bond Elut Plexa PAX Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Bond Elut Plexa PAX nonpolar/anion-exchange SPE is suitable for polar and nonpolar acidic analytes.
Expand 5 Items
Loading...
Bond Elut Certify Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Bond Elut Certify is a mixed-mode phase ideal for extraction of basic drugs from urine and blood.
Expand 3 Items
Loading...
Bond Elut Plexa PCX Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Bond Elut Plexa PCX Plates, Agilent Technologies
Expand 3 Items
Loading...
Bond Elut PBA Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Bond Elut PBA uses phenylboronic acid functionality to retain analytes.
Expand 1 Items
Loading...
Sort by
Recommended for You