Order Entry
Canada
ContactUsLinkComponent
Zoology/Ornithology Storage Trays
Zoology/Ornithology Storage Trays
Catalog # 470022-342
Supplier:  Lane Science
Zoology/Ornithology Storage Trays
Catalog # 470022-342
Supplier:  Lane Science
Supplier Number:  201 1/2 T

Specifications

  • Description:
    Zoology/Ornithology Storage Trays
  • Cat. no.:
    470022-342
  • Supplier No.:
    201 1/2 T

Specifications

About this item

Aluminum frames and masonite bottoms make drawers sturdy, while plastic runners make them easy to slide in and out of the cabinet.

  • Size: 42³/₈×26×1⁵/₈" Ext.Ø each
  • Weight: 9 lbs., 8 oz.

Each drawer also features built-in label holders.