ContactUsLinkComponent
Flint Glass Y-Shaped Connecting Tubes
Catalog # 470235-774
Supplier:  GSC International
undefined
Flint Glass Y-Shaped Connecting Tubes
Catalog # 470235-774
Supplier:  GSC International
Supplier Number:  2004-4-10
Specifications
  • Ext.Ø:
    5/16"
  • Ø int.:
    1/4"
  • Description:
    Flint Glass Y-Shaped Connecting Tubes, Arm Length: 1 1/2"
  • Material:
    Flint glass
  • Cat. no.:
    470235-774
Specifications
About this item
Y-shaped connector with a 120° angle provides safer connections as the ends have constrictions to hold rubber tubing securely.
  • Made from quality flint glass