Order Entry
Canada
Orders LinkContactUsLinkComponent
Xylene

Xylene

Supplier:   Ward's Science
Xylene
Supplier:  Ward's Science
Certificates
Xylene

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 1330-20-7
Formula Weight: 106.17
Formula: C6H4(CH3)2
Density (g/mL): 0.87
Boiling Point (°C): 140
Shelf Life (months): 36
Storage: Red

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.

Product Family Options

Product Information

  • Grade
    Size
    Chemical/physical specifications
    Availability
    Price
  • Xylene
    Reagent
    500 mL
    Solution
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $42.35
    $42.35
    Each (500ml)
       $42.35

    Supplier Number: XX0020-500MLR
  • Xylene
    Laboratory
    500 mL
    Solution
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $41.40
    $41.40
    Each (500ml)
       $41.40

    Supplier Number: XX0020-500ML

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.