Orders LinkContactUsLinkComponent
Wire Triangles
Wire Triangles
Supplier:   GSC International
Wire Triangles
Supplier:  GSC International
Certificates
Wire Triangles
3 Options Available Below
Product Details & Documents
Perfect tool for suspending a crucible.
  • Uniform clay tubes
  • Galvanized iron wire resists rusting
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • 470235-758
    Side length: 38 mm
    Side length: 38 mm
    20 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $42.65
    $42.65
    Log in for availability
    Case of 100
    Case of 100 - $387.75
    $387.75
    Pack of 10
       $42.65
    Case of 100
       $387.75
    Supplier Number: 4-2019-CS
  • 470235-760
    Side length: 51 mm
    Side length: 51 mm
    26 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $43.75
    $43.75
    Log in for availability
    Case of 100
    Case of 100 - $383.45
    $383.45
    Pack of 10
       $43.75
    Case of 100
       $383.45
    Supplier Number: 4-2020-CS
  • 470235-762
    Side length: 64 mm
    Side length: 64 mm
    33 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $47.85
    $47.85
    Log in for availability
    Case of 100
    Case of 100 - $414.90
    $414.90
    Pack of 10
       $47.85
    Case of 100
       $414.90
    Supplier Number: 4-2030-CS