Wide Range pH Test Paper
pH Test Paper, pH 1 - 14
Catalog # 470203-388
Supplier: Micro Essential
Supplier #: 94
Jump to:
Product Details & Documents
Features Sharp Color Change.
- Sensitive to the entire pH range 1-14
- Color comparator chart provided with each dispenser
- 4.6 m roll
Specifications
- Description:pH Test Paper
- pH range:pH 1 - 14



