ContactUsLinkComponent
Water Quality Testing III Kit: Multiplexed PCR for Testing Water Bacteria
Water Quality Testing III Kit: Multiplexed PCR for Testing Water Bacteria
Catalog # 470120-914
Supplier:  Edvotek
undefined
Water Quality Testing III Kit: Multiplexed PCR for Testing Water Bacteria
Catalog # 470120-914
Supplier:  Edvotek
Supplier Number:  953
Specifications
  • Description:
    Water Quality Testing III Kit: Multiplexed PCR for Testing Water Bacteria
  • No. of Lab Groups:
    10
  • Cat. no.:
    470120-914
  • Supplier No.:
    953
Specifications
About this item
Students check water for contaminants using PCR-based testing.
  • Kit Uses Nonhazardous Microorganisms
  • Students Practice Lab Procedures
  • Instructions Included
  • PCR Used in a Multiplexed Format
  • Material for Ten Labs with 20 Students Each
Students perform screening procedures for waterborne bacteria in drinking water. Using a single PCR reaction, they test for the presence of three separate and classroom-safe microorganisms.
The kit has enough material for ten experiments with 20 students.
Ordering information: This kit includes primer mixtures; 200 base pair ladder; control DNAs; non-pathogenic E. coli, B. subtilis, and S. marcescens; chelating agent; proteinase k; PCR beads (taq polymerase, dNTPS, buffer); ultrapure H20; TE buffer; 10X gel loading solution; InstaStain Ethidium Bromide; InstaStain Methylene Blue; micro test tubes with attached caps; and instructions with background information. It requires but does not include thermal cycler, water baths, or heating baths for PCR; horizontal gel electrophoresis apparatus; DC power supply; micropipettors with tips; balance; 500 mL flask; microcentrifuge; UV transilluminator or white light; and a microwave.