Orders LinkContactUsLinkComponent
Wall Mounted Eyewash Station
Wall Mounted Eyewash Station
Wall Mounted Eyewash
Catalog # 470205-492
Supplier:   WaterSaver Faucet
Supplier #:  EW814
Wall Mounted Eyewash Station
Catalog # 470205-492
Supplier:  WaterSaver Faucet
Wall Mounted Eyewash Station
Wall Mounted Eyewash Station
Wall Mounted Eyewash
Price
$1,093.60
Log in for availability
Each
Product Details & Documents
This wall-mounted unit quickly and gently rinses contaminates away from both eyes simultaneously.
  • Stainless steel bowl for safe, simple use
  • Two heads deliver flood of water for hands-free eye rinsing
  • Heavy-duty pipe and fittings resist corrosion
Featuring two GS-Plus™ (Gentle Spray) Heads, this low-density polyethylene filter removes scale, deposits, and debris from the water and keeps water line impurities out of users' eyes.
A push handle activates the eye wash, which allows for hands free operation.
Certifications: ANSI-complaint identification sign included.
Specifications
  • Description:
    Wall Mounted Eyewash