Orders LinkContactUsLinkComponent
Standard-Grade Glass Beaker
Standard-Grade Glass Beaker
Standard-Grade Glass Beaker
Supplier:  VWR International
Certificates
Standard-Grade Glass Beaker
Standard-Grade Glass Beaker
12 Options Available Below
Product Details & Documents
Our high-quality borosilicate beakers meet ASTM standards and are a great value. With uniform wall thickness, white enamel, double-graduated metric scale, and a marking spot. Beakers can be heated and used with acids, salt solutions, and organic solvents.
Documents
Product Family Options
Product Information
  • Capacity
    Graduation Interval
    Graduation Range
    Availability
    Price
  • 10 ml (0.34 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $8.85
    $8.85
    Log in for availability
    Case of 48
    Case of 48 - $375.90
    $375.90
    Each
       $8.85
    Case of 48
       $375.90
  • 20 ml (0.68 oz.)
    5 ml
    5 - 15 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $8.85
    $8.85
    Log in for availability
    Case of 48
    Case of 48 - $409.55
    $409.55
    Each
       $8.85
    Case of 48
       $409.55
  • 30 ml
    10 ml
    5 - 25 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $9.15
    $9.15
    Log in for availability
    Case of 48
    Case of 48 - $398.15
    $398.15
    Each
       $9.15
    Case of 48
       $398.15
  • 50 ml
    10 ml
    10 - 40 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $10.90
    $10.90
    Log in for availability
    Case of 48
    Case of 48 - $463.90
    $463.90
    Each
       $10.90
    Case of 48
       $463.90
  • 100 ml (3.4 oz.)
    10 ml
    20 - 80 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $10.60
    $10.60
    Log in for availability
    Case of 48
    Case of 48 - $465.80
    $465.80
    Each
       $10.60
    Case of 48
       $465.80
  • 470191-148
    Standard-grade glass beaker, 150 ml
    Standard-grade glass beaker, 150 ml
    10 ml
    20 - 140 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $11.20
    $11.20
    Log in for availability
    Case of 48
    Case of 48 - $476.35
    $476.35
    Each
       $11.20
    Case of 48
       $476.35
  • 250 ml
    25 ml
    25 - 200 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $11.20
    $11.20
    Log in for availability
    Case of 48
    Case of 48 - $487.45
    $487.45
    Each
       $11.20
    Case of 48
       $487.45
  • 400 ml
    25 ml
    25 - 325 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $11.70
    $11.70
    Log in for availability
    Case of 48
    Case of 48 - $501.80
    $501.80
    Each
       $11.70
    Case of 48
       $501.80
  • 470191-152
    Standard-grade glass beaker, 600 ml
    Standard-grade glass beaker, 600 ml
    50 ml
    50 - 500 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $12.50
    $12.50
    Log in for availability
    Case of 36
    Case of 36 - $396.40
    $396.40
    Each
       $12.50
    Case of 36
       $396.40
  • 800 ml
    50 ml
    50 - 750 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $12.00
    $12.00
    Log in for availability
    Case of 24
    Case of 24 - $255.50
    $255.50
    Each
       $12.00
    Case of 24
       $255.50
  • 1000 ml
    100 ml
    50 - 1000 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $27.20
    $27.20
    Log in for availability
    Case of 24
    Case of 24 - $589.35
    $589.35
    Each
       $27.20
    Case of 24
       $589.35
  • 2000 ml
    100 ml
    200 - 1800 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $41.70
    $41.70
    Log in for availability
    Case of 8
    Case of 8 - $294.85
    $294.85
    Each
       $41.70
    Case of 8
       $294.85