Order Entry
Canada
ContactUsLinkComponent
VWR® One Block Heater
VWR® One Block Heater
Catalog # 470204-396
Supplier:  VWR International
undefined
VWR® One Block Heater
Catalog # 470204-396
Supplier:  VWR International

Specifications

  • Description:
    Block for 20, 1.5 mL Tubes
  • Cat. no.:
    470204-396
  • Supplier No.:
    949010

Specifications

About this item

One Block Heaters are ideal for a variety of molecular biology procedures such as enzyme reactions, enzyme deactivations, and culture incubation.

  • Temperature Range: Ambient +5 To 150°C
  • VAC: 120
  • Average Weight: 6 1/10 lb.
  • Length: 12½"
  • Width: 8"
  • Height: 3½"

The versatile heaters weigh 6 1/10 lb.

The temperature is fully adjustable, and the counter displays the heating time.