ContactUsLinkComponent
VWR® Berzelius Tall Form Beakers
  470191-230
undefined
VWR® Berzelius Tall Form Beakers
  470191-230
Select an Option
1000 ml (34 oz.)
Select an Option
470191-230
Select an Option
V1060-1000
Our new high-quality borosilicate Berzelius beakers meet ASTM standard and are a great value.
  • Borosilicate glass fabrication
  • Used with acids, salt solutions, and organic solvents
Each has a marking spot and may be used with acids, salt solutions, and organic solvents.
Frequently Bought Together