ContactUsLinkComponent
Polymethylpentene (Nalgene) Volumetric Flasks with Caps
Polymethylpentene (Nalgene) Volumetric Flasks with Caps
  470211-822
undefined
Polymethylpentene (Nalgene) Volumetric Flasks with Caps
  470211-822
 :  4001-1000
  • Capacity:
    1000 ml
  • Cat. no.:
    470211-822
  • Supplier No.:
    4001-1000
Polymethylpentene flasks are highly resistant to most reagents. They have the clarity and accuracy of glass without the fragility.
All flasks are calibrated with the same precision of high quality glass and can be autoclaved. A leak-proof polypropylene screw cap is included with each.