Orders LinkContactUsLinkComponent
Vernier® Voltage Probe
Vernier® Voltage Probe
Vernier voltage probe (BTA), ±10 V
Catalog # CA470017-076
New Product
Supplier #:  VP-BTA
Vernier® Voltage Probe
Catalog # CA470017-076
New Product
Vernier® Voltage Probe
Vernier® Voltage Probe
Vernier voltage probe (BTA), ±10 V
Price
$27.09
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The Vernier Voltage Probe is a convenient and easy way to measure voltage.
  • Easy-To-Use
  • Convenient
  • Applicable in Physics, Chemistry, Physical Science, or Middle School Science Classes
  • Purpose in Physics Classes: to Measure Potential in Direct or Alternating-Current Circuits
  • Purpose in Science Classes: to Measure Voltages Developed in Varieties of Electrochemical or Voltaic Cells
Measure direct voltage with the sensor’s red test clips.
In physics classes, the probe can be used to measure the potential in direct-current or alternating-current circuits. In chemistry, physical science, or middle school science classes, students measure voltages developed in a variety of electrochemical or voltaic cells.
Specifications
  • Description:
    Vernier voltage probe (BTA)
  • Range:
    ±10 V