ContactUsLinkComponent
Biocam Virus Chart Set
Biocam Virus Chart Set
Catalog # 470023-544
undefined
Biocam Virus Chart Set
Catalog # 470023-544
Supplier Number:  SPECIAL
Specifications
  • Width:
    45.7 cm
  • Description:
    Virus Chart Set
  • Length:
    61 cm
  • Cat. no.:
    470023-544
  • Finish:
    Laminated
  • Supplier No.:
    SPECIAL
Specifications
About this item
Comprehensive Overview of Structure and Replication
  • Colorful illustrations and descriptive text
These charts present useful information for your virology studies. The Virus I chart depicts well-known examples of icosahedral, helical, enveloped, and complex virus structures; Virus II chart covers replication, with detail on Influenza life cycle; Virus III presents information specific to bacteriophages, contrasting lytic and lysogenic cycles. Set of three charts.