Vernier® Salinity Sensor
About this item
Easily and precisely measure the total dissolved salt content in ocean or brackish water.
Response time: 90% of full-scale reading in 10 s.
2 Options Available Below
Product Details & Documents
Easily and precisely measure the total dissolved salt content in ocean or brackish water.
Response time: 90% of full-scale reading in 10 s.
Specifications for Product Family
Specifications
- Catalog No:
- CA470017-558
- CA470219-326
- Salinity range:
- 0 - 50 ppt (0 - 50,000 ppm)
- Salinity accuracy:
- Using factory calibration: ±3% of full-scale reading, Using custom calibration: ±1% of full-scale reading
- Temperature range:
- 0...80 °C
- Dimensions:
- 12 OD×150L mm
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- CA470017-558Vernier salinity sensor (BTA)Supplier Number: SAL-BTASpecifications:New Product
More
Specifications
Cat. No.CA470017-558Salinity range0 - 50 ppt (0 - 50,000 ppm)Salinity accuracyUsing factory calibration: ±3% of full-scale reading, Using custom calibration: ±1% of full-scale readingTemperature range0...80 °CDimensions12 OD×150L mmNew Product - CA470219-326Vernire salinitysStandard 35 ppt, 500 mlSupplier Number: SAL-STSpecifications:New Product
More
Specifications
Cat. No.CA470219-326Salinity rangeSalinity accuracyTemperature rangeDimensionsNew Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



