Vernier® pH Sensors
About this item
General-purpose pH Sensors can be used across disciplines, with applications in chemistry, biology, middle school science, and environmental science.
- Automated Data Collection
- Graphing
- Data Analysis
- For Titrations, Studies of Household Acids and Bases, and More
- Ideal for Use in Multiple Disciplines
Advantages of these sensors include automated data collection, graphing, and data analysis.
Typical activities using pH Sensors include acid-base titrations, studies of household acids and bases, monitoring pH change during chemical reactions or in an aquarium as a result of photosynthesis, investigations of acid rain and buffering, and analysis of water quality in streams and lakes.
2 Options Available Below
Product Details & Documents
General-purpose pH Sensors can be used across disciplines, with applications in chemistry, biology, middle school science, and environmental science.
- Automated Data Collection
- Graphing
- Data Analysis
- For Titrations, Studies of Household Acids and Bases, and More
- Ideal for Use in Multiple Disciplines
Advantages of these sensors include automated data collection, graphing, and data analysis.
Typical activities using pH Sensors include acid-base titrations, studies of household acids and bases, monitoring pH change during chemical reactions or in an aquarium as a result of photosynthesis, investigations of acid rain and buffering, and analysis of water quality in streams and lakes.
Documents
Vernier pH Sensor.pdf
Manual
Specifications for Product Family
Specifications
- Catalog No:
- CA470039-294
- CA470219-320
- Response time:
- 90% of final reading in 1 second
- Accuracy:
- ±0.2 pH units
- Operating temperature range:
- 5...80°C
Specifications
Product Family Options
Product Information
- ElectrolyteAvailabilityPrice
- CA470039-294Vernier pH sensor (BTA)Supplier Number: PH-BTASpecifications:New Product
More
Specifications
Cat. No.CA470039-294Response time90% of final reading in 1 secondAccuracy±0.2 pH unitsOperating temperature range5...80°CNew Product - CA470219-320Vernier pH storage solution, 500 mlSupplier Number: PH-SSSpecifications:New Product
More
Specifications
Cat. No.CA470219-320Response timeAccuracyOperating temperature rangeNew Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.