Vernier® ORP Sensor
Vernier ORP sensor (BTA)
Catalog # CA470017-890
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Supplier #: ORP-BTA
Jump to:
Product Details & Documents
Measure the ability of a solution to act as an oxidizing agent or reducing agent with this oxidation-reduction potential sensor.
Positive readings indicate an oxidizing agent, while negative readings indicate a reducing agent. ORP electrodes are often used to measure the oxidizing ability of chlorine in swimming pools, or to determine when the equivalence point has been reached in an oxidation-reduction reaction.
Range: -450 to +1100 mV.
Specifications
- Description:Vernier ORP sensor (BTA)