Vernier® Electrode Amplifier
Product Details & Documents
The Electrode Amplifier is an ORP/mV/pH amplifier that accepts an electrode with a standard BNC connector. It amplifies a –450 mV to +1100 mV range to the 0 - 5 V range of our interfaces.
- Power: 7 mA at 5 VDC
- Input range: –450 mV to +1100 mV
- Impedance: >100 MΩ
- Gain: 2.2 V/V and offset: 1.20 V (nominal)
The electrode amplifier is an ORP/mV/pH amplifier that accepts an electrode with a standard BNC connector. It amplifies a -450 mV to +1100 mV range to the 0-5 V range of our interfaces. It can be used with any of the following: replacement pH electrode, most third-party pH electrodes (with BNC connectors), other high output impedance eletrodes. Most third-party ORP electrodes (with BNC connectors).
Specifications
- Description:Electrode amplifier (BTA)
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



