Orders LinkContactUsLinkComponent
Utility Shears
Utility Shears
  470007-988
 :  27-755-020
Utility Shears
  470007-988
Utility Shears
Utility Shears
Utility Shears
$18.95
Each Case of 200
Cut everything from paper to 24-gauge stainless steel with these shears.
  • Lightweight, 420 stainless steel blades (Rockwell hardness of 500)
  • Autoclavable polypropylene handles
Length: 9"
  • Description:
    Utility Shears