Urinary Bladder, Collapsed Slide
Catalog # 470182-934
Supplier: Avantor
Specifications
- Description:Urinary Bladder, Collapsed Slide
- Cat. no.:470182-934
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-934
- Orientation:Section
Specifications
About this item
Component of the urinary system
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Demonstrates transitional epithelium, lamina propria, and muscularis of a collasped bladder.