ContactUsLinkComponent
Universal Clamp with Clamp Holder
Universal Clamp with Clamp Holder
Catalog # 470112-748
undefined
Universal Clamp with Clamp Holder
Catalog # 470112-748
Supplier Number:  ECBH01
Specifications
  • Description:
    Universal Clamp with Clamp Holder
  • Cat. no.:
    470112-748
  • Jaw Type:
    3-prong
  • Jaw Width:
    57 mm
  • Supplier No.:
    ECBH01
Specifications
About this item
Great for Securing Glassware.
  • Maximum jaw opening 2 1/4"
  • Vinyl-coated jaws and fiberglass sleeves protect glassware
Jaws and extension rod are rust resistant alloy. Distance between apparatus and support rod is adjustable. A ribbed, thumbscrew clamp holder allows jaw position to be rotated 360°.