ContactUsLinkComponent
Ungraduated Pyrex® Centrifuge Tubes
Ungraduated Pyrex® Centrifuge Tubes
Catalog # 470211-616
Supplier:  Corning
undefined
Ungraduated Pyrex® Centrifuge Tubes
Catalog # 470211-616
Supplier:  Corning
Supplier Number:  8060-15
Specifications
  • Capacity:
    15 ml
  • Cat. no.:
    470211-616
  • Supplier No.:
    8060-15
Specifications
About this item
Made of PYREX® borosilicate glass, conical, beaded.
  • Wide neck
  • Excellent chemical resistance
  • Excellent temperature resistance
These centrifuge tubes has uniform glass distribution in the wall, taper and bottom provides improved mechanical strength, prevents the formation of failure-causing stress regions during centrifuging and insures the highest degree of accuracy.