Orders LinkContactUsLinkComponent
U-Form Electromagnet
U-Form Electromagnet
U-Form Electromagnet
Catalog # 470120-696
Supplier:   Wards
Supplier #:  PH0800B
U-Form Electromagnet
Catalog # 470120-696
Supplier:  Wards
U-Form Electromagnet
U-Form Electromagnet
U-Form Electromagnet
Clearance
$19.85
Sale
Valid until 31/12/2026
Log in for availability
Each
Product Details & Documents
Quality electromagnet.
  • Wound with insulated copper wire
  • Provided with barrel connections
This unique electromagnet consists of an iron U-shaped core with armature and a carrying hook. The electromagnet is designed for use on 4–6 volts DC.
Specifications
  • Description:
    U-Form Electromagnet