Assortment of Two-Hole Rubber Stoppers
Catalog # 470123-296
Supplier: United Scientific Supplies
Specifications
- Description:Two-Hole Rubber Stoppers, Assorted Sizes
- Cat. no.:470123-296
- Supplier No.:RSTPK4
Specifications
About this item
One Pound of Two-Hole Rubber Stoppers in Various Sizes.
- Pliable
- Autoclavable
- Sizes: #2 to #8
Black stoppers are made of solid natural rubber that stays pliable over a long period of time. Ideal for use with non-aggressive or concentrated solutions. Stoppers feature a low sulfur content, and will withstand temperatures from –25 to 70 °C (–13 to 158 °F).