Orders LinkContactUsLinkComponent
Fruit and Potato Clock
Fruit and Potato Clock
Two Potato Clock
Catalog # 470006-220
Supplier:   Wards
Supplier #:  PT-CLK
Fruit and Potato Clock
Catalog # 470006-220
Supplier:  Wards
Fruit and Potato Clock
Fruit and Potato Clock
Two Potato Clock
Price
$41.90
Log in for availability
Each
Product Details & Documents
This LCD clock keeps time using potatoes, soda, or plants.
  • Length: 8½"
  • Width: 3½"
  • Height: 2¾"
  • Demonstration of an Electrochemical Cell
  • Instructions Included
It's an ideal demonstration of an electrochemical cell. All you need to do is attach the included zinc and copper bimetallic probes and watch the resettable hours, minutes, seconds, and date light up.
The clock includes instructions and measures 2¾ x 3½ x 8½".
Specifications
  • Description:
    Two Potato Clock