Triple Sugar Iron (TSI) Agar Tubes
Catalog # 470180-650
Supplier: Avantor
CAS Number:
Specifications
- Description:Prepared TSI Agar Tubes
- Cat. no.:470180-650
- Supplier No.:WARD470180-650
Specifications
About this item
For identification of Gram-negative bacteria based on dextrose, lactose, and sucrose fermentation, and hydrogen sulfide production.
Frequently Bought Together