ContactUsLinkComponent
T-Shaped Flint Glass Connecting Tubes
T-Shaped Flint Glass Connecting Tubes
Catalog # 470235-772
Supplier:  GSC International
undefined
T-Shaped Flint Glass Connecting Tubes
Catalog # 470235-772
Supplier:  GSC International
Supplier Number:  2003-1-10
Specifications
  • Ext.Ø:
    5/16"
  • Ø int.:
    1/4"
  • Description:
    T-Shaped Flint Glass Connecting Tubes, Arm Length: 1 1/2"
  • Material:
    Flint glass
  • Cat. no.:
    470235-772
Specifications
About this item
For joining two rubber tubes to a common tube at right angles. Safer connections are provided as the ends have constrictions to hold rubber tubing securely.
  • Made from quality flint glass