How Clean is your Lab Station? Microbiology Experiment Kit
470105-940
- Description:Microbiology Experiment Kit
- Cat. no.:470105-940
- Supplier No.:CM100-B200
Test for the presence of bacteria in your workspace before and after cleaning with this TSA Microbiology Experiment Kit.
- Four popular experiments
Using the included materials, students can conduct a variety of bacteria tests to discover the characteristics of bacteria and conditions under which it is most prevalent.
The unique set up of this kit allows students to melt the media in a microwave, pour their own media plates, and test for bacteria growth in four popular experiments.
Kit Includes:
Two 200ml bottles of prepared Tryptic Soy Agar
20 gamma radiated sterile Petri dishes
10 sterile swabs
Instruction sheets.
: This item will ship immediately upon order, unless you specify otherwise.