Orders LinkContactUsLinkComponent
Tripod Stands
Tripod Stands
Tripod Stands, Inside Diameter: 4 1/2", 6"
Catalog # 470123-750
Supplier #:  TRS6X9
Tripod Stands
Catalog # 470123-750
Tripod Stands
Tripod Stands
Tripod Stands, Inside Diameter: 4 1/2", 6"
Price
$40.75
Log in for availability
Each
Product Details & Documents
A must have for every lab.
  • Cast iron, enameled finish
  • Removable zinc plated legs
Good quality, enameled cast iron tripods for supporting small objects over a burner. The legs detach for easy storage.
Specifications
  • Ext.Ø:
    6"
  • Description:
    Tripod Stands, Inside Diameter: 4 1/2"