Orders LinkContactUsLinkComponent
Tripod Ring Stands
Tripod Ring Stands
Tripod Ring Stand, 3 1/2" dia., One Ring
Catalog # 470019-528
Supplier #:  TRS4X9
Tripod Ring Stands
Catalog # 470019-528
Tripod Ring Stands
Tripod Ring Stands
Tripod Ring Stand, 3 1/2" dia., One Ring
Price
$41.25
Log in for availability
Each
Product Details & Documents
Support stand for holding labware over a burner.
  • Cast iron ring
  • Detachable legs
  • Zinc plated legs
Hold beakers and flasks at a safe distance from the heat source such as a bunsen burner using these tripods. Each comes standard with one ring and detachable legs. Size: 9"H.
Specifications
  • Description:
    Tripod Ring Stand, 3 1/2" dia., One Ring