Orders LinkContactUsLinkComponent
Tri Pour Beakers
Tri Pour Beakers
Supplier:   VWR International
Tri Pour Beakers
Supplier:  VWR International
Certificates
Tri Pour Beakers
Tri Pour Beakers
5 Options Available Below
Product Details & Documents
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
  • Accurate graduations to ±5
  • Tri-cornered lip for dripless pouring
  • Unbreakable
  • Resistant to common acids, solvents, and alkalis
  • Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 470174-588
    Tri-pour beakers
    Tri-pour beakers
    50 ml (1.7 oz.)
    Discontinued
  • 470174-586
    Tri-pour beakers
    Tri-pour beakers
    100 ml (3.4 oz.)
    Discontinued
  • 490012-442
    Tri-Pour Beakers
    Tri-Pour Beakers
    100 ml (3.4 oz.)
    Discontinued
  • 470174-582
    Tri-pour beakers
    Tri-pour beakers
    400 ml (14 oz.)
    Discontinued
  • 470174-590
    Tri-Pour Beakers, Set of 6 different sizes
    Tri-Pour Beakers, Set of 6 different sizes
    50 ml, 100 ml, 250 ml, 400 ml, 800 ml, 1000 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $20.75
    $20.75
    Log in for availability
    Case of 30
    Case of 30 - $94.20
    $94.20
    Pack of 6
       $20.75
    Case of 30
       $94.20
  • 470361-522
    Tri-pour beakers
    Tri-pour beakers
    50 ml (1.7 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1.26
    $1.26
    Log in for availability
    Case of 100
    Case of 100 - $126.40
    $126.40
    Each
       $1.26
    Case of 100
       $126.40
    Specifications:
    New Product
    New Product
  • 470174-587
    Tri-pour beakers
    Tri-pour beakers
    100 ml (3.4 oz.)
    Discontinued
    Specifications:
    New Product
    New Product
  • 470174-583
    Tri-pour beakers
    Tri-pour beakers
    400 ml (14 oz.)
    Discontinued
    Specifications:
    New Product
    New Product
  • 470361-520
    Tri-pour beakers
    Tri-pour beakers
    1 L (34 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $3.24
    $3.24
    Log in for availability
    Case of 100
    Case of 100 - $323.90
    $323.90
    Each
       $3.24
    Case of 100
       $323.90
    Specifications:
    New Product
    New Product
  • 470303-163
    Tri-pour beakers
    Tri-pour beakers
    100 ml (3.4 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1.34
    $1.34
    Log in for availability
    Case of 100
    Case of 100 - $134.30
    $134.30
    Each
       $1.34
    Case of 100
       $134.30
    Specifications:
    New Product
    New Product
  • 470303-161
    Tri-pour beakers
    Tri-pour beakers
    400 ml (14 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $2.21
    $2.21
    Log in for availability
    Case of 100
    Case of 100 - $221.20
    $221.20
    Each
       $2.21
    Case of 100
       $221.20
    Specifications:
    New Product