Order Entry
Canada
ContactUsLinkComponent
 
Ward's® Tree Of Life Chart
  470177-214
undefined
Ward's® Tree Of Life Chart
  470177-214
 :  7192600

 

  • Width:
    140 cm
  • Description:
    Tree Of Life
  • Length:
    152 cm
  • Cat. no.:
    470177-214
  • Finish:
    Unlaminated
  • Supplier No.:
    7192600

 

 

Classification Of Living Things In A Simplified Presentation

  • Helps students gain a better perspective on the inter-relationships of living things

Follow the separate branches to locate the different types of life-forms, as well as examples of each.